Gene Transcripts

Gene Symbol
CLLU1OS
Entrez Gene ID
574016
Gene Name
chronic lymphocytic leukemia up-regulated 1 opposite strand
Gene Aliases
Chromosome Location
Chr. 12: 92420086 - 92428449 on Build GRCh38
UniGene ID
Hs.551199
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001025232.1 NP_001020403.1 4 471
XM_017019673.1 XP_016875162.1 4 757
GenBank mRNA AJ845168.1 - 4 471

View Full Product Details