Gene Transcripts

Gene Symbol
FMN1
Entrez Gene ID
342184
Gene Name
formin 1
Gene Aliases
FMN, LD
Chromosome Location
Chr. 15: 32765544 - 33194765 on Build GRCh38
UniGene ID
Hs.657649
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001277313.1 NP_001264242.1 4 614
NM_001277314.1 NP_001264243.1 4 614
XM_011521504.2 XP_011519806.1 4 437
XM_011521505.2 XP_011519807.1 4 646
XM_011521506.2 XP_011519808.1 4 645
XM_011521507.2 XP_011519809.1 4 646
XM_017022130.1 XP_016877619.1 1 808
XM_017022131.1 XP_016877620.1 4 646
XM_017022133.1 XP_016877622.1 4 643
XM_017022135.1 XP_016877624.1 4 643
GenBank mRNA CR749487.1 - 4 578

View Full Product Details