Gene Transcripts
- Gene Symbol
- TMEM14B
- Entrez Gene ID
- 81853
- Gene Name
- transmembrane protein 14B
- Gene Aliases
- Chromosome Location
- Chr. 6: 10747759 - 10759774 on Build GRCh38
- UniGene ID
- Hs.667371
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001127711.2 |
NP_001121183.1
|
3, 4 |
276 |
|
NM_001286484.1 |
NP_001273413.1
|
3, 4 |
301 |
|
NM_001286488.1 |
NP_001273417.1
|
4, 5 |
378 |
|
NM_001286489.1 |
NP_001273418.1
|
3, 4 |
276 |
|
NM_030969.4 |
NP_112231.3
|
4, 5 |
378 |
|
GenBank mRNA |
AK291883.1 |
- |
4, 5 |
332 |
|
AK294152.1 |
- |
4, 5 |
332 |
|
BC001033.1 |
- |
4, 5 |
320 |
3139565 |
BC007080.1 |
- |
4, 5 |
332 |
4066217 |
BC013913.1 |
- |
4, 5 |
340 |
3685635 |
BC071660.1 |
- |
4, 5 |
322 |
4473732 |
BG288654.1 |
- |
3, 4 |
230 |
|
BG432075.1 |
- |
4, 5 |
307 |
|
BP249229.1 |
- |
3, 4 |
230 |
|
BQ056988.1 |
- |
3, 4 |
229 |
|
View Full Product Details