Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_000971.3 | NP_000962.2 | 1 | 142 | |
XM_006716463.2 | XP_006716526.1 | 2, 3 | 735 | ||
GenBank mRNA | AK225246.1 | - | 1 | 141 | |
AK225254.1 | - | 1 | 140 | ||
AK291119.1 | - | 2, 3 | 140 | ||
AK313365.1 | - | 2, 3 | 140 | ||
BC006095.1 | - | 1 | 130 | 2821364 | |
BC008850.2 | - | 1 | 167 | 4138113 | |
BC009599.1 | - | 1 | 142 | 4134557 | |
BC071671.1 | - | 1 | 123 | 4392472 | |
BC071894.1 | - | 1 | 127 | 2906310 | |
BC071895.1 | - | 1 | 125 | 6568654 | |
BC087837.1 | - | 1 | 126 | 6170371 | |
CR456773.1 | - | 1 | 120 | ||
L16558.1 | - | 1 | 120 | ||
X52967.1 | - | 2, 3 | 141 | ||
X57958.1 | - | 1 | 157 | ||
X57959.1 | - | 1 | 130 |