Gene Transcripts
- Gene Symbol
- MCHR2-AS1
- Entrez Gene ID
- 728012
- Gene Name
- MCHR2 antisense RNA 1
- Gene Aliases
- Chromosome Location
- Chr. 6: 99994038 - 100076419 on Build GRCh38
- UniGene ID
- Hs.434366
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
DB461069.1 |
- |
2 |
294 |
|
View Full Product Details