Gene Transcripts

Gene Symbol
C12orf42
Entrez Gene ID
374470
Gene Name
chromosome 12 open reading frame 42
Gene Aliases
Chromosome Location
Chr. 12: 103047669 - 103525258 on Build GRCh38
UniGene ID
Hs.534649
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_011538312.2 XP_011536614.1 10 629

View Full Product Details