Gene Transcripts
- Gene Symbol
- ASB9P1
- Entrez Gene ID
- 728619
- Gene Name
- ankyrin repeat and SOCS box containing 9 pseudogene 1
- Gene Aliases
- Chromosome Location
- Chr. 15: 92795484 - 92797108 on Build GRCh38
- UniGene ID
- Hs.684129
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_033769.1 |
|
1 |
415 |
|
View Full Product Details