Gene Transcripts

Gene Symbol
KIF27
Entrez Gene ID
55582
Gene Name
kinesin family member 27
Gene Aliases
Chromosome Location
Chr. 9: 83832795 - 83921539 on Build GRCh38
UniGene ID
Hs.697514
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_011518850.2 XP_011517152.1 1 80
XM_011518857.2 XP_011517159.1 1 80
XM_017014902.1 XP_016870391.1 1 80

View Full Product Details