Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_000175.3 | NP_000166.2 | 14 | 1398 | |
NM_001184722.1 | NP_001171651.1 | 14 | 1461 | ||
NM_001289789.1 | NP_001276718.1 | 15 | 1545 | ||
NM_001289790.1 | NP_001276719.1 | 13 | 1314 | ||
XM_005258764.2 | XP_005258821.1 | 15 | 1364 | ||
XM_006723148.2 | XP_006723211.1 | 15 | 1315 | ||
XM_011526754.2 | XP_011525056.1 | 16 | 1780 | ||
GenBank mRNA | AK025421.1 | - | 3 | 996 | |
AK129884.1 | - | 15 | 1329 | ||
AK130102.1 | - | 13 | 1093 | ||
AK293446.1 | - | 13 | 1190 | ||
AK294396.1 | - | 14 | 1409 | ||
AK301103.1 | - | 14 | 1202 | ||
BC004982.1 | - | 14 | 1266 | 2906270 | |
K03515.1 | - | 14 | 1273 | ||
X16539.1 | - | 14 | 1264 |