Gene Transcripts

Gene Symbol
FEZF1
Entrez Gene ID
389549
Gene Name
FEZ family zinc finger 1
Gene Aliases
FEZ, HH22, ZNF312B
Chromosome Location
Chr. 7: 122300939 - 122311855 on Build GRCh38
UniGene ID
Hs.553970
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_005250337.3 XP_005250394.1 1 116
XM_011516202.2 XP_011514504.1 1 116

View Full Product Details