Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001512.3 | NP_001503.1 | 7 | 724 | |
XM_005249035.3 | XP_005249092.1 | 7 | 817 | ||
XM_011514534.2 | XP_011512836.1 | 6 | 667 | ||
XM_011514535.2 | XP_011512837.1 | 6 | 763 | ||
GenBank mRNA | AF020918.1 | - | 7 | 644 | |
AF025887.1 | - | 7 | 647 | ||
AF125271.1 | - | 6 | 574 | ||
AF125272.1 | - | 6 | 574 | ||
AF125273.1 | - | 6 | 574 | ||
AK054804.1 | - | 5 | 1081 | ||
AK315369.1 | - | 7 | 623 | ||
BC015523.2 | - | 6 | 594 | 3915425 | |
BC063439.1 | - | 5 | 561 | 5248427 | |
CR749474.1 | - | 3 | 4023 | ||
Y13047.1 | - | 6 | 574 |