Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_015732.4 | NP_056547.3 | 2 | 500 | |
GenBank mRNA | AF073788.1 | - | 2 | 365 | |
AF205889.1 | - | 1 | 205 | ||
AK049192.1 | - | 2 | 444 | ||
AK142333.1 | - | 2 | 502 | ||
BC057338.1 | - | 2 | 483 | 6827741 |