Gene Transcripts

Gene Symbol
Runx2
Entrez Gene ID
12393
Gene Name
runt related transcription factor 2
Gene Aliases
AML3, Cbf, Cbfa-1, Cbfa1, LS3, Osf2, PEBP2aA, Pebp2a1, Pebpa2a
Chromosome Location
Chr. 17: 44495986 - 44814831 on Build GRCm38
UniGene ID
Mm.391013
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001145920.2 NP_001139392.1 7 2355
NM_001146038.2 NP_001139510.1 9 1620
NM_001271627.1 NP_001258556.1 8 1784
NM_001271630.1 NP_001258559.1 6 1709
NM_001271631.1 NP_001258560.1 5 1604
NM_009820.5 NP_033950.2 9 1587
GenBank mRNA AF005936.1 - 8 1738
AF010284.1 - 9 1587
AK089291.1 - 5 1140
D14636.1 - 7 2353
DQ458792.1 - 8 1372

View Full Product Details