Gene Transcripts

Gene Symbol
Cbfa2t3
Entrez Gene ID
12398
Gene Name
core-binding factor, runt domain, alpha subunit 2, translocated to, 3 (human)
Gene Aliases
A630044F12Rik, AI465270, AW229127, Cbfa2t3h, ETO-2, Eto2, MTGR2
Chromosome Location
Chr. 8: 122625136 - 122699639 on Build GRCm38
UniGene ID
Mm.194339
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001109873.1 NP_001103343.1 5 854
NM_009824.2 NP_033954.2 5 835
NM_177289.1 NP_796263.1 4 776
GenBank mRNA AF038029.1 - 5 717
AK033680.1 - 4 673
AK041887.1 - 5 856
BB658300.1 - 5 597
BC141396.1 - 5 664 9056009
BC151156.1 - 5 664 9088044

View Full Product Details