Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_009949.2 | NP_034079.2 | 4 | 1642 | |
GenBank mRNA | AK144441.1 | - | 4 | 1642 | |
AK150163.1 | - | 4 | 1614 | ||
AK169038.1 | - | 4 | 1539 | ||
BC138514.1 | - | 4 | 1577 | 8861534 | |
BC145859.1 | - | 4 | 1577 | 40131162 | |
U01163.1 | - | 4 | 1587 |