Gene Transcripts

Gene Symbol
Cyp2a12
Entrez Gene ID
13085
Gene Name
cytochrome P450, family 2, subfamily a, polypeptide 12
Gene Aliases
Chromosome Location
Chr. 7: 27029090 - 27036818 on Build GRCm38
UniGene ID
Mm.20770
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_133657.1 NP_598418.1 4 633
GenBank mRNA BC018356.1 - 4 633
BC094017.1 - 4 621 5101743
L06463.1 - 4 619

View Full Product Details