Gene Transcripts

Gene Symbol
Evi5
Entrez Gene ID
14020
Gene Name
ecotropic viral integration site 5
Gene Aliases
NB4S
Chromosome Location
Chr. 5: 107744795 - 107875107 on Build GRCm38
UniGene ID
Mm.35796
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_007964.2 NP_031990.2 3 373
GenBank mRNA AK134294.1 - 3 574
AK162905.1 - 3 633
AK165332.1 - 4 366
AK165885.1 - 3 326
AK216641.1 - 1 49
BC030056.1 - 4 380
BI737814.1 - 3 297
U53586.1 - 3 373

View Full Product Details