Gene Transcripts
- Gene Symbol
- Il4i1
- Entrez Gene ID
- 14204
- Gene Name
- interleukin 4 induced 1
- Gene Aliases
- Fig1, Fig1-ps, LAAO, LAO
- Chromosome Location
- Chr. 7: 44836288 - 44840809 on Build GRCm38
- UniGene ID
- Mm.2565
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC115960.1 |
- |
8 |
928 |
|
U70429.1 |
- |
8 |
940 |
|
View Full Product Details