Gene Transcripts

Gene Symbol
Flt3
Entrez Gene ID
14255
Gene Name
FMS-like tyrosine kinase 3
Gene Aliases
B230315G04, CD135, Flk-2, Flk2, Flt-3, Ly72, wmfl
Chromosome Location
Chr. 5: 147330741 - 147400489 on Build GRCm38
UniGene ID
Mm.194
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010229.2 NP_034359.2 8 1200
GenBank mRNA AK045865.1 - 8 1023
AK149292.1 - 8 1018
AK163640.1 - 8 1201
BC109003.1 - 8 993
BC109004.1 - 8 1005
M64689.1 - 8 1006
X59398.1 - 8 1057

View Full Product Details