Gene Transcripts

Gene Symbol
Gdf10
Entrez Gene ID
14560
Gene Name
growth differentiation factor 10
Gene Aliases
Bmp3b
Chromosome Location
Chr. 14: 33923587 - 33935285 on Build GRCm38
UniGene ID
Mm.432071
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145741.2 NP_665684.2 2 1022
GenBank mRNA AK042684.1 - 1013
BC022669.1 - 1 332
BC058358.1 - 2 1022 6828166
L42114.1 - 2 838
S82648.1 - 2 838

View Full Product Details