Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001167694.2 | NP_001161166.1 | 4 | 577 | |
NM_001167695.2 | NP_001161167.1 | 4 | 508 | ||
NM_001167696.2 | NP_001161168.1 | 4 | 538 | ||
NM_001167697.2 | NP_001161169.1 | 3 | 625 | ||
NM_001167699.2 | NP_001161171.1 | 2 | 507 | ||
NM_001167700.2 | NP_001161172.1 | 5 | 727 | ||
NM_008157.3 | NP_032183.2 | 2 | 468 | ||
GenBank mRNA | AA041917.1 | - | 2 | 404 | |
AK132917.1 | - | 5 | 666 | ||
AK140770.1 | - | 5 | 923 | ||
BC021648.1 | - | 4 | 516 | 3989077 | |
BC050187.1 | - | 3 | 596 | 30020126 | |
DV660907.1 | - | 4 | 511 | ||
U46923.1 | - | 2 | 328 |