Gene Transcripts

Gene Symbol
Guca1a
Entrez Gene ID
14913
Gene Name
guanylate cyclase activator 1a (retina)
Gene Aliases
GC-A, Gcap1, Guca1, mGCAP1
Chromosome Location
Chr. 17: 47394558 - 47413514 on Build GRCm38
UniGene ID
Mm.16224
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_008189.2 NP_032215.2 1 346
GenBank mRNA BC026834.1 - 1 346
BC031810.1 - 1 313 4501021
L36860.1 - 1 203

View Full Product Details