Gene Transcripts

Gene Symbol
H2-T3
Entrez Gene ID
15043
Gene Name
histocompatibility 2, T region locus 3
Gene Aliases
H-2T3, H2-Tw3, TL
Chromosome Location
Chr. 17: 36185571 - 36190151 on Build GRCm38
UniGene ID
Mm.426002
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_008208.4 NP_032234.3 2 233
GenBank mRNA AK033602.1 - 2 234
BC125378.1 - 2 231 40129893
BC125380.1 - 2 231 40129895
D86082.1 - 2 210

Gene Symbol
H2-T3-like
Entrez Gene ID
547348
Gene Name
MHC class I antigen
Gene Aliases
Chromosome Location
Chr. : - on Build
UniGene ID
Mm.426002
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AY989873.1 - 1 383

View Full Product Details