Gene Transcripts

Gene Symbol
Hexb
Entrez Gene ID
15212
Gene Name
hexosaminidase B
Gene Aliases
Chromosome Location
Chr. 13: 97176331 - 97198357 on Build GRCm38
UniGene ID
Mm.27816
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010422.2 NP_034552.1 12 1524
GenBank mRNA AK151149.1 - 12 1461
AK159134.1 - 12 1477
AK159178.1 - 12 1498
AK160560.1 - 12 1493
AK168825.1 - 12 1498
U07633.1 - 12 1496
Y00964.1 - 12 1437

View Full Product Details