Gene Transcripts

Gene Symbol
Hmx3
Entrez Gene ID
15373
Gene Name
H6 homeobox 3
Gene Aliases
Nkx-5.1, Nkx5-1, Nkx5.1
Chromosome Location
Chr. 7: 131541138 - 131546027 on Build GRCm38
UniGene ID
Mm.323562
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_008257.3 NP_032283.3 2 736
GenBank mRNA BC139482.1 - 2 832
BC139483.1 - 2 832 8862505

View Full Product Details