Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001012306.2 | NP_001012306.2 | 3 | 820 | |
NM_001161742.1 | NP_001155214.1 | 5 | 1099 | ||
NM_001161743.1 | NP_001155215.1 | 5 | 1046 | ||
NM_001161744.1 | NP_001155216.1 | 4 | 1038 | ||
NM_001161745.1 | NP_001155217.1 | 4 | 985 | ||
GenBank mRNA | BC089581.1 | - | 3 | 820 | |
M77015.1 | - | 4 | 911 |