Gene Transcripts

Gene Symbol
HOXC13
Entrez Gene ID
3229
Gene Name
homeobox C13
Gene Aliases
ECTD9, HOX3, HOX3G
Chromosome Location
Chr. 12: 53938792 - 53946544 on Build GRCh38
UniGene ID
Hs.118608
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017410.2 NP_059106.2 2 1722
GenBank mRNA AF255676.1 - 2 1718
AF263466.2 - 2 1678
AK024027.1 - 2 1204
BC002754.1 - 3 1622
BC090850.1 - 2 1691 6171228

View Full Product Details