Gene Transcripts

Gene Symbol
HOXD9
Entrez Gene ID
3235
Gene Name
homeobox D9
Gene Aliases
HOX4, HOX4C, Hox-4.3, Hox-5.2
Chromosome Location
Chr. 2: 176122685 - 176124917 on Build GRCh38
UniGene ID
Hs.236646
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_014213.3 NP_055028.3 1 847
GenBank mRNA BC044855.1 - 1 847
X59372.1 - 1 769

View Full Product Details