Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_013594.2 | NP_038622.2 | 15 | 1879 | |
GenBank mRNA | AF072240.1 | - | 17 | 2026 | |
AK004624.2 | - | 15 | 1880 | ||
AK156401.1 | - | 16 | 1870 | ||
BC069837.1 | - | 15 | 1895 | 30435760 |