Gene Transcripts

Gene Symbol
PGRMC2
Entrez Gene ID
10424
Gene Name
progesterone receptor membrane component 2
Gene Aliases
DG6, PMBP
Chromosome Location
Chr. 4: 128269237 - 128288829 on Build GRCh38
UniGene ID
Hs.507910
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_006320.4 NP_006311.2 3 1657
GenBank mRNA AJ002030.1 - 3 624
AK091741.1 - 4 594
AK094949.1 - 2 744
AK131272.1 - 3 1657
BC016692.1 - 3 641 4067832
BC092478.1 - 3 672 4831185

View Full Product Details