Gene Transcripts
- Gene Symbol
- Klk1b4
- Entrez Gene ID
- 18048
- Gene Name
- kallikrein 1-related pepidase b4
- Gene Aliases
- Ngfa, mGk-4
- Chromosome Location
- Chr. 7: 44207435 - 44211754 on Build GRCm38
- UniGene ID
- Mm.440886
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
M11434.1 |
- |
|
555 |
|
View Full Product Details