Gene Transcripts
- Gene Symbol
- Cntn3
- Entrez Gene ID
- 18488
- Gene Name
- contactin 3
- Gene Aliases
- Pang
- Chromosome Location
- Chr. 6: 102163304 - 102564620 on Build GRCm38
- UniGene ID
- Mm.435479
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_008779.2 |
NP_032805.2
|
8 |
1156 |
|
GenBank mRNA |
L01991.1 |
- |
|
1265 |
|
View Full Product Details