Gene Transcripts
- Gene Symbol
- Ppt1
- Entrez Gene ID
- 19063
- Gene Name
- palmitoyl-protein thioesterase 1
- Gene Aliases
- 9530043G02Rik, AA960502, C77813, CLN1, D4Ertd184e, INCL, PPT
- Chromosome Location
- Chr. 4: 122836075 - 122859175 on Build GRCm38
- UniGene ID
- Mm.277719
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_008917.3 |
NP_032943.2
|
4, 5 |
463 |
|
GenBank mRNA |
AF071025.1 |
- |
4, 5 |
480 |
|
AF087568.1 |
- |
4, 5 |
441 |
|
AF326557.1 |
- |
4, 5 |
447 |
|
AF326558.1 |
- |
4, 5 |
447 |
|
AK014561.1 |
- |
4, 5 |
445 |
|
AK148314.1 |
- |
4, 5 |
438 |
|
AK151055.1 |
- |
4, 5 |
438 |
|
AK151107.1 |
- |
4, 5 |
449 |
|
AK152474.1 |
- |
4, 5 |
447 |
|
AK152940.1 |
- |
4, 5 |
449 |
|
AK153106.1 |
- |
4, 5 |
444 |
|
AK153316.1 |
- |
4, 5 |
444 |
|
AK169579.1 |
- |
5, 6 |
538 |
|
AK169658.1 |
- |
5, 6 |
544 |
|
AK171668.1 |
- |
4, 5 |
445 |
|
AK192077.1 |
- |
2, 3 |
118 |
|
AK192572.1 |
- |
2, 3 |
118 |
|
AK216167.1 |
- |
2, 3 |
115 |
|
View Full Product Details