Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_031874.4 | NP_114080.2 | 2 | 422 | |
GenBank mRNA | AB232590.1 | - | 1 | 188 | |
AF263365.1 | - | 2 | 422 | ||
AK048395.1 | - | 2 | 362 | ||
AK078462.1 | - | 2 | 301 | ||
AK169369.1 | - | 2 | 300 | ||
BC010779.1 | - | 2 | 289 | 4216156 | |
BC020010.1 | - | 2 | 314 | 3584902 | |
M89777.1 | - | 1 | 231 |