Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001166409.2 | NP_001159881.1 | 2 | 316 | |
NM_001166410.2 | NP_001159882.1 | 2 | 316 | ||
NM_001166411.2 | NP_001159883.1 | 2 | 316 | ||
NM_001293658.1 | NP_001280587.1 | 2 | 316 | ||
NM_016809.6 | NP_058089.2 | 2 | 316 | ||
GenBank mRNA | AB016424.1 | - | 2 | 143 | |
AK003720.1 | - | 2 | 134 | ||
AK007354.1 | - | 2 | 134 | ||
AK049575.1 | - | 2 | 134 | ||
AK088031.1 | - | 2 | 132 | ||
AK133795.1 | - | 2 | 136 | ||
AK154044.1 | - | 2 | 138 | ||
AK167800.1 | - | 2 | 132 | ||
AK167967.1 | - | 2 | 136 | ||
AK168285.1 | - | 2 | 137 | ||
AK168338.1 | - | 2 | 136 | ||
BC006580.1 | - | 2 | 111 | 3586577 | |
BC059098.1 | - | 2 | 122 | 4013810 | |
BC086491.1 | - | 2 | 127 | 30434553 | |
BC087737.1 | - | 1 | 82 | ||
BC106176.1 | - | 2 | 172 | 30146829 | |
CF912757.1 | - | 2 | 316 | ||
CN696660.1 | - | 2 | 149 | ||
DV045913.1 | - | 2 | 151 |