Gene Transcripts

Gene Symbol
Ren1
Entrez Gene ID
19701
Gene Name
renin 1 structural
Gene Aliases
D19352, Ren, Ren-1, Ren-A, Ren1c, Ren1d, Rn-1, Rnr
Chromosome Location
Chr. 1: 133350674 - 133360320 on Build GRCm38
UniGene ID
Mm.220955
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031192.3 NP_112469.1 7, 8 986
GenBank mRNA AK052685.1 - 7, 8 986
AK085309.1 - 7, 8 986
BC061053.1 - 7, 8 984 30313659
X16642.1 - 7, 8 968

View Full Product Details