Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_009059.2 | NP_033085.2 | 13 | 1636 | |
GenBank mRNA | AK144440.1 | - | 13 | 1638 | |
BC068121.1 | - | 13 | 1636 | 6415928 | |
KJ650210.1 | - | 12 | 1444 | ||
U54639.1 | - | 13 | 1565 |