Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_009143.3 | NP_033169.3 | 3 | 698 | |
GenBank mRNA | AK002496.1 | - | 3 | 454 | |
BC058798.1 | - | 3 | 698 | 6485217 | |
BC062881.1 | - | 3 | 444 | 5693309 | |
BC087871.1 | - | 3 | 343 | 30279332 | |
D50646.1 | - | 3 | 452 |