Gene Transcripts

Gene Symbol
Sh2b1
Entrez Gene ID
20399
Gene Name
SH2B adaptor protein 1
Gene Aliases
AI425885, C530001K22Rik, Irip, Psm, SH2-B, SH2-Bb, Sh2bpsm1, mKIAA1299
Chromosome Location
Chr. 7: 126466993 - 126475684 on Build GRCm38
UniGene ID
Mm.8538
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001081459.2 NP_001074928.1 2 1330
NM_001289538.1 NP_001276467.1 2 1330
NM_001289539.1 NP_001276468.1 2 953
NM_001289540.1 NP_001276469.1 3 1090
NM_001289541.1 NP_001276470.1 2 953
NM_001289542.1 NP_001276471.1 2 1330
NM_011363.3 NP_035493.2 2 1330
GenBank mRNA AF020526.1 - 2 1024
AF074329.1 - 1 735
AF380422.1 - 2 1024
AF421138.1 - 2 1024
AF421139.1 - 2 1024
AK148085.1 - 3 1057
AK154655.1 - 2 1264
AK168439.1 - 2 925
AK170444.1 - 2 1304
AK171328.1 - 2 1333
AK173145.1 - 1 477
AK187869.1 - 1 169
AK191125.1 - 1 169
BC011422.1 - 1 792 3156603
BC051978.1 - 2 873 6401205

View Full Product Details