Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001289724.1 | NP_001276653.1 | 4 | 704 | |
NM_001289725.1 | NP_001276654.1 | 4 | 659 | ||
NM_009193.2 | NP_033219.1 | 5 | 764 | ||
GenBank mRNA | AK131729.1 | - | 535 | ||
AK136165.1 | - | 1 | 54 | ||
AK154053.1 | - | 5 | 511 | ||
AK162121.1 | - | 5 | 510 | ||
BC025113.1 | - | 5 | 498 | ||
BC029631.1 | - | 4 | 436 | 3500676 | |
U75680.1 | - | 5 | 496 |