Gene Transcripts

Gene Symbol
FOXK2
Entrez Gene ID
3607
Gene Name
forkhead box K2
Gene Aliases
ILF, ILF-1, ILF1
Chromosome Location
Chr. 17: 82519718 - 82604607 on Build GRCh38
UniGene ID
Hs.591140
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_004514.3 NP_004505.2 8 1846
GenBank mRNA U58196.1 - 8 1857
U58197.1 - 8 1857

View Full Product Details