Gene Transcripts

Gene Symbol
Snn
Entrez Gene ID
20621
Gene Name
stannin
Gene Aliases
2810407J07Rik, AI848521, AW547286
Chromosome Location
Chr. 16: 11066298 - 11074985 on Build GRCm38
UniGene ID
Mm.325800
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_009223.3 NP_033249.1 2 769
GenBank mRNA AK149685.1 - 2 718
BC006961.1 - 2 737 3707801
BC017685.1 - 2 686 4241981
BC038475.1 - 1 810 3157762
BC063027.1 - 2 706 5343626
BC087932.1 - 714 6706156

View Full Product Details