Gene Transcripts

Gene Symbol
Tmie
Entrez Gene ID
20776
Gene Name
transmembrane inner ear
Gene Aliases
5131400L21Rik, Mm.87012, sr
Chromosome Location
Chr. 9: 110865707 - 110880083 on Build GRCm38
UniGene ID
Mm.390229
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_146260.2 NP_666372.1 4 1363
GenBank mRNA AF481143.1 - 4 1363
AK030341.1 - 4 1393
AK031966.1 - 4 1425
BC138840.1 - 4 927 8861862
BC141124.1 - 4 927 9055736

View Full Product Details