Gene Transcripts

Gene Symbol
Sycp1
Entrez Gene ID
20957
Gene Name
synaptonemal complex protein 1
Gene Aliases
SCP1
Chromosome Location
Chr. 3: 102818499 - 102936312 on Build GRCm38
UniGene ID
Mm.243849
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_011516.2 NP_035646.2 24, 25 2103
GenBank mRNA AK076780.1 - 3, 4 193
AK133229.1 - 24, 25 2103
BC137967.1 - 24, 25 2065 8860986
D88539.1 - 19, 20 1693
L41069.1 - 24, 25 2084
Z38118.1 - 24, 25 2094

View Full Product Details