Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_011535.3 | NP_035665.2 | 2 | 1420 | |
NM_198052.2 | NP_932169.1 | 3 | 1616 | ||
GenBank mRNA | AF429310.1 | - | 2 | 274 | |
AK052453.1 | - | 2 | 1635 | ||
AK145436.1 | - | 2 | 1337 | ||
AK157247.1 | - | 2 | 1181 | ||
AK172118.1 | - | 2 | 1183 | ||
BC096551.1 | - | 3 | 1616 | 30547736 | |
U15567.1 | - | 1 | 85 | ||
U57328.1 | - | 2 | 301 |