Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_009402.2 | NP_033428.1 | 2 | 308 | |
GenBank mRNA | AF076482.1 | - | 2 | 307 | |
AF193843.1 | - | 2 | 296 | ||
AK008335.1 | - | 2 | 309 | ||
BC005582.1 | - | 2 | 318 | 3969014 | |
CT010165.1 | - | 2 | 272 | ||
X86374.1 | - | 2 | 307 |