Gene Transcripts

Gene Symbol
Tpst1
Entrez Gene ID
22021
Gene Name
protein-tyrosine sulfotransferase 1
Gene Aliases
R75054, Tango13a
Chromosome Location
Chr. 5: 130073326 - 130135734 on Build GRCm38
UniGene ID
Mm.332720
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001130476.2 NP_001123948.1 2 603
NM_013837.2 NP_038865.1 2 420
GenBank mRNA AF038008.1 - 2 420
AK150753.1 - 2 488
BC012666.1 - 2 350 4035106
CJ046784.1 - 2 604
CT010202.1 - 1 210

View Full Product Details