Gene Transcripts

Gene Symbol
Ttr
Entrez Gene ID
22139
Gene Name
transthyretin
Gene Aliases
AA408768, AI787086, D17860, prealbumin
Chromosome Location
Chr. 18: 20665250 - 20674326 on Build GRCm38
UniGene ID
Mm.2108
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_013697.5 NP_038725.1 3 477
GenBank mRNA AK014454.1 - 3 314
AK018701.1 - 3 315
AK050155.1 - 3 314
AK075588.1 - 3 314
BC024702.1 - 3 447 4192268
BC032069.1 - 3 304
BC086926.1 - 3 314 6749152
CK627038.1 - 3 299
D89076.1 - 3 314
X03351.1 - 3 314

View Full Product Details