Gene Transcripts

Gene Symbol
Vip
Entrez Gene ID
22353
Gene Name
vasoactive intestinal polypeptide
Gene Aliases
Chromosome Location
Chr. 10: 5639193 - 5647617 on Build GRCm38
UniGene ID
Mm.98916
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_011702.2 NP_035832.1 4 425
GenBank mRNA AK018599.1 - 4 426
AK043544.1 - 3 2146
BC089511.1 - 4 331 30249277

View Full Product Details