Gene Transcripts

Gene Symbol
Wbp4
Entrez Gene ID
22380
Gene Name
WW domain binding protein 4
Gene Aliases
AW545037, BB101031, FBP21
Chromosome Location
Chr. 14: 79459937 - 79481268 on Build GRCm38
UniGene ID
Mm.476813
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_018765.3 NP_061235.2 3, 4 472
GenBank mRNA AF071184.1 - 3, 4 324
AK019160.1 - 3, 4 206
AK135215.1 - 3, 4 472
AK159522.1 - 3, 4 724
AK160559.1 - 3, 4 209
AK161496.1 - 3, 4 166
AK162009.1 - 3, 4 214
BC034851.1 - 3, 4 167 1399984

View Full Product Details